Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGTGTGAGTGGCTTTAGTAAG[C/T]AATTCATGTAATGACATTTTTCCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF222 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF222 - zinc finger protein 222 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129996.1 | Intron | NP_001123468.1 | ||||
NM_013360.2 | Intron | NP_037492.2 | ||||
XM_005259208.3 | Intron | XP_005259265.1 | ||||
XM_005259209.3 | Intron | XP_005259266.1 | ||||
XM_017027237.1 | Intron | XP_016882726.1 | ||||
XM_017027238.1 | Intron | XP_016882727.1 |
Set Membership: |
HapMap |