Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCCCACATACATCAATTTCTGG[C/T]TCTTATTTTTTATTATTTTTAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606191 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FNBP1 - formin binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR107 - G protein-coupled receptor 107 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136557.1 | Intron | NP_001130029.1 | ||||
NM_001136558.1 | Intron | NP_001130030.1 | ||||
NM_001287346.1 | Intron | NP_001274275.1 | ||||
NM_020960.4 | Intron | NP_066011.2 | ||||
XM_017014953.1 | Intron | XP_016870442.1 | ||||
XM_017014954.1 | Intron | XP_016870443.1 |
Set Membership: |
HapMap |