Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCACATGCAGTGACTCTCCCTT[C/T]TGGGTTTCCACTGAAGGACCTTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 153420 | ||||||||||||||||||||
Literature Links: |
CD58 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD58 - CD58 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144822.1 | 12848 | Intron | NP_001138294.1 | |||
NM_001779.2 | 12848 | Intron | NP_001770.1 | |||
XM_017002869.1 | 12848 | UTR 3 | XP_016858358.1 |
LOC105378922 - uncharacterized LOC105378922 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |