Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCGGGTTCTTGGGCTGGCCGTG[A/T]CCTTCTCCCTCGGTCGCCACTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM260 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM260 - transmembrane protein 260 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017799.3 | 63 | UTR 5 | NP_060269.3 | |||
XM_005267771.1 | 63 | Intron | XP_005267828.1 | |||
XM_006720176.1 | 63 | UTR 5 | XP_006720239.1 | |||
XM_006720178.1 | 63 | UTR 5 | XP_006720241.1 | |||
XM_011536851.1 | 63 | UTR 5 | XP_011535153.1 | |||
XM_011536852.1 | 63 | UTR 5 | XP_011535154.1 | |||
XM_011536853.1 | 63 | UTR 5 | XP_011535155.1 | |||
XM_017021379.1 | 63 | UTR 5 | XP_016876868.1 | |||
XM_017021380.1 | 63 | UTR 5 | XP_016876869.1 |