Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATCAGATTAAGACATTTGGGTAA[A/C]ATAATAGACCCTGAACTAGAGGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610585 MIM: 601158 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP22 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGAP22 - Rho GTPase activating protein 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256024.1 | Intron | NP_001242953.1 | ||||
NM_001256025.2 | Intron | NP_001242954.1 | ||||
NM_001256026.1 | Intron | NP_001242955.1 | ||||
NM_021226.3 | Intron | NP_067049.2 | ||||
XM_005270014.3 | Intron | XP_005270071.1 | ||||
XM_011540002.1 | Intron | XP_011538304.1 | ||||
XM_011540003.1 | Intron | XP_011538305.1 | ||||
XM_011540004.1 | Intron | XP_011538306.1 | ||||
XM_011540005.1 | Intron | XP_011538307.1 | ||||
XM_011540006.1 | Intron | XP_011538308.1 | ||||
XM_011540007.2 | Intron | XP_011538309.1 | ||||
XM_011540011.2 | Intron | XP_011538313.1 | ||||
XM_011540012.2 | Intron | XP_011538314.1 | ||||
XM_011540013.2 | Intron | XP_011538315.1 | ||||
XM_011540015.2 | Intron | XP_011538317.1 | ||||
XM_017016470.1 | Intron | XP_016871959.1 | ||||
XM_017016471.1 | Intron | XP_016871960.1 | ||||
XM_017016472.1 | Intron | XP_016871961.1 | ||||
XM_017016473.1 | Intron | XP_016871962.1 | ||||
XM_017016474.1 | Intron | XP_016871963.1 | ||||
XM_017016475.1 | Intron | XP_016871964.1 | ||||
XM_017016476.1 | Intron | XP_016871965.1 | ||||
XM_017016477.1 | Intron | XP_016871966.1 |
MAPK8 - mitogen-activated protein kinase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278547.1 | Intron | NP_001265476.1 | ||||
NM_001278548.1 | Intron | NP_001265477.1 | ||||
NM_001323302.1 | Intron | NP_001310231.1 | ||||
NM_001323320.1 | Intron | NP_001310249.1 | ||||
NM_001323321.1 | Intron | NP_001310250.1 | ||||
NM_001323322.1 | Intron | NP_001310251.1 | ||||
NM_001323323.1 | Intron | NP_001310252.1 | ||||
NM_001323324.1 | Intron | NP_001310253.1 | ||||
NM_001323325.1 | Intron | NP_001310254.1 | ||||
NM_001323326.1 | Intron | NP_001310255.1 | ||||
NM_001323327.1 | Intron | NP_001310256.1 | ||||
NM_001323328.1 | Intron | NP_001310257.1 | ||||
NM_001323329.1 | Intron | NP_001310258.1 | ||||
NM_001323330.1 | Intron | NP_001310259.1 | ||||
NM_001323331.1 | Intron | NP_001310260.1 | ||||
NM_139046.3 | Intron | NP_620634.1 | ||||
NM_139049.3 | Intron | NP_620637.1 |
Set Membership: |
HapMap |