Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGCTAGGAGGTAACTCTTTGG[C/T]CTACATCGAATGTCATGATACCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607315 MIM: 603602 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CABP5 - calcium binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019855.4 | Intron | NP_062829.1 | ||||
XM_017026992.1 | Intron | XP_016882481.1 |
PLA2G4C - phospholipase A2 group IVC | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159322.1 | Intron | NP_001152794.1 | ||||
NM_001159323.1 | Intron | NP_001152795.1 | ||||
NM_003706.2 | Intron | NP_003697.2 | ||||
XM_011527431.2 | Intron | XP_011525733.1 | ||||
XM_011527432.2 | Intron | XP_011525734.1 | ||||
XM_011527433.2 | Intron | XP_011525735.1 | ||||
XM_017027411.1 | Intron | XP_016882900.1 | ||||
XM_017027412.1 | Intron | XP_016882901.1 | ||||
XM_017027413.1 | Intron | XP_016882902.1 | ||||
XM_017027414.1 | Intron | XP_016882903.1 |