Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACATGGATGCCAGAAACTTCCAT[C/G]GACCATACACCTACAAACATGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612404 | ||||||||||||||||||||
Literature Links: |
RASL11B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RASL11B - RAS like family 11 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCFD2 - sec1 family domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152540.3 | 3127 | UTR 3 | NP_689753.2 | |||
XM_011534375.2 | 3127 | Intron | XP_011532677.1 | |||
XM_011534376.2 | 3127 | Intron | XP_011532678.1 | |||
XM_011534378.2 | 3127 | Intron | XP_011532680.1 | |||
XM_017007786.1 | 3127 | Intron | XP_016863275.1 | |||
XM_017007787.1 | 3127 | Intron | XP_016863276.1 |