Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGAACAAAGCCGATGACAGCCG[G/T]ATGAGCTTGTTGGGAAAACTGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf26 - chromosome 7 open reading frame 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303039.1 | 611 | Silent Mutation | CGG,CGT | R,R 91 | NP_001289968.1 | |
NM_024067.3 | 611 | Silent Mutation | CGG,CGT | R,R 110 | NP_076972.2 | |
XM_005249845.4 | 611 | Silent Mutation | CGG,CGT | R,R 110 | XP_005249902.1 | |
XM_011515518.2 | 611 | Silent Mutation | CGG,CGT | R,R 110 | XP_011513820.1 | |
XM_017012614.1 | 611 | UTR 5 | XP_016868103.1 | |||
XM_017012615.1 | 611 | UTR 5 | XP_016868104.1 |
ZDHHC4 - zinc finger DHHC-type containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |