Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAGGACTCACGGGTCCTGGGCA[C/G]CAGCATTGACTTTGGATGGTTGGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608869 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100996251 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100996251 - uncharacterized LOC100996251 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRIG2 - leucine rich repeats and immunoglobulin like domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312686.1 | Intron | NP_001299615.1 | ||||
NM_014813.2 | Intron | NP_055628.1 | ||||
XM_005271369.2 | Intron | XP_005271426.1 | ||||
XM_017002952.1 | Intron | XP_016858441.1 |
Set Membership: |
HapMap |