Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615850 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS53 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VPS53 - VPS53, GARP complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128159.2 | 10474 | UTR 3 | NP_001121631.1 | |||
NM_018289.3 | 10474 | Intron | NP_060759.2 | |||
XM_011523953.2 | 10474 | Intron | XP_011522255.1 | |||
XM_017024817.1 | 10474 | Intron | XP_016880306.1 | |||
XM_017024818.1 | 10474 | Intron | XP_016880307.1 |