Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605199 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFPQ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SFPQ - splicing factor proline and glutamine rich | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005066.2 | 7809 | Intron | NP_005057.1 | |||
XM_005271112.4 | 7809 | Intron | XP_005271169.1 | |||
XM_005271113.4 | 7809 | Intron | XP_005271170.1 | |||
XM_005271115.4 | 7809 | Intron | XP_005271172.1 | |||
XM_011541950.2 | 7809 | UTR 3 | XP_011540252.1 | |||
XM_017002053.1 | 7809 | UTR 3 | XP_016857542.1 | |||
XM_017002054.1 | 7809 | UTR 3 | XP_016857543.1 |
Set Membership: |
HapMap |