Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609245 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPSM2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPSM2 - G-protein signaling modulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321038.1 | 989 | Intron | NP_001307967.1 | |||
NM_001321039.1 | 989 | Intron | NP_001307968.1 | |||
NM_013296.4 | 989 | Intron | NP_037428.3 | |||
XM_006710589.2 | 989 | Intron | XP_006710652.1 | |||
XM_011541301.2 | 989 | Intron | XP_011539603.1 | |||
XM_011541302.2 | 989 | Intron | XP_011539604.1 | |||
XM_011541303.2 | 989 | Intron | XP_011539605.1 | |||
XM_017001097.1 | 989 | UTR 5 | XP_016856586.1 | |||
XM_017001098.1 | 989 | UTR 5 | XP_016856587.1 |