Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTACATTAAGTAATCTATTCTAG[A/G]AAGCCACAAACTGGGGAACAACAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615900 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGBL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AGBL5 - ATP/GTP binding protein like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035507.2 | Intron | NP_001030584.1 | ||||
NM_021831.5 | Intron | NP_068603.4 | ||||
XM_005264477.2 | Intron | XP_005264534.1 | ||||
XM_011533011.2 | Intron | XP_011531313.1 | ||||
XM_011533012.2 | Intron | XP_011531314.1 | ||||
XM_011533013.2 | Intron | XP_011531315.1 |
AGBL5-AS1 - AGBL5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRA-AGC8-1 - transfer RNA-Ala (AGC) 8-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |