Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAACTGTAGGAGATTATTTTGAA[G/T]TAGTTGAAAATAATGTTGGAGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBXN2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UBXN2A - UBX domain protein 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181713.3 | Intron | NP_859064.2 | ||||
XM_005264166.4 | Intron | XP_005264223.1 | ||||
XM_005264167.4 | Intron | XP_005264224.1 | ||||
XM_005264168.4 | Intron | XP_005264225.1 | ||||
XM_005264169.4 | Intron | XP_005264226.1 | ||||
XM_011532633.2 | Intron | XP_011530935.1 | ||||
XM_017003517.1 | Intron | XP_016859006.1 | ||||
XM_017003518.1 | Intron | XP_016859007.1 |
Set Membership: |
HapMap |