Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGAAGGACACTTTGCTCATTGCA[A/G]AGTGATCAGGAGACACCATTAGTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C1QTNF7 - C1q and tumor necrosis factor related protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135170.1 | Intron | NP_001128642.1 | ||||
NM_001135171.1 | Intron | NP_001128643.1 | ||||
NM_031911.4 | Intron | NP_114117.1 | ||||
XM_011513772.1 | Intron | XP_011512074.1 | ||||
XM_011513773.1 | Intron | XP_011512075.1 |
LOC101929095 - uncharacterized LOC101929095 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |