Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604354 | ||||||||||||||||||||
Literature Links: |
GPALPP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPALPP1 - GPALPP motifs containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316951.1 | 195 | Nonsense Mutation | GAG,TAG | E,* 24 | NP_001303880.1 | |
NM_001316952.1 | 195 | UTR 5 | NP_001303881.1 | |||
NM_018559.3 | 195 | Nonsense Mutation | GAG,TAG | E,* 24 | NP_061029.2 | |
XM_005266444.3 | 195 | Nonsense Mutation | GAG,TAG | E,* 24 | XP_005266501.1 | |
XM_011535144.2 | 195 | UTR 5 | XP_011533446.1 |
NUFIP1 - NUFIP1, FMR1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012345.2 | 195 | Intron | NP_036477.2 |