Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM207A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM207A - family with sequence similarity 207 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316983.1 | Intron | NP_001303912.1 | ||||
NM_001316984.1 | Intron | NP_001303913.1 | ||||
NM_001316985.1 | Intron | NP_001303914.1 | ||||
NM_001316986.1 | Intron | NP_001303915.1 | ||||
NM_001316987.1 | Intron | NP_001303916.1 | ||||
NM_001316988.1 | Intron | NP_001303917.1 | ||||
NM_058190.3 | Intron | NP_478070.1 | ||||
XM_017028481.1 | Intron | XP_016883970.1 | ||||
XM_017028482.1 | Intron | XP_016883971.1 |
LINC01547 - long intergenic non-protein coding RNA 1547 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |