Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300142 | ||||||||||||||||||||
Literature Links: |
PAK3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAK3 - p21 (RAC1) activated kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128166.2 | Intron | NP_001121638.1 | ||||
NM_001128167.2 | Intron | NP_001121639.1 | ||||
NM_001128168.2 | Intron | NP_001121640.1 | ||||
NM_001128172.2 | Intron | NP_001121644.1 | ||||
NM_001128173.2 | Intron | NP_001121645.1 | ||||
NM_001324325.1 | Intron | NP_001311254.1 | ||||
NM_001324326.1 | Intron | NP_001311255.1 | ||||
NM_001324327.1 | Intron | NP_001311256.1 | ||||
NM_001324328.1 | Intron | NP_001311257.1 | ||||
NM_001324329.1 | Intron | NP_001311258.1 | ||||
NM_001324330.1 | Intron | NP_001311259.1 | ||||
NM_001324331.1 | Intron | NP_001311260.1 | ||||
NM_001324332.1 | Intron | NP_001311261.1 | ||||
NM_001324333.1 | Intron | NP_001311262.1 | ||||
NM_001324334.1 | Intron | NP_001311263.1 | ||||
NM_002578.4 | Intron | NP_002569.1 |