Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAATGAGGACCCAGGCCCCTGGG[A/G]GAGTACAGAATTCCAGCCAGGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191305 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BLK - BLK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001715.2 | Intron | NP_001706.2 | ||||
XM_011543824.1 | Intron | XP_011542126.1 | ||||
XM_011543825.2 | Intron | XP_011542127.1 | ||||
XM_011543827.1 | Intron | XP_011542129.1 | ||||
XM_011543828.2 | Intron | XP_011542130.1 | ||||
XM_011543829.2 | Intron | XP_011542131.1 | ||||
XM_017013730.1 | Intron | XP_016869219.1 |
Set Membership: |
HapMap |