Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610949 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SYT14 - synaptotagmin 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146261.2 | Intron | NP_001139733.1 | ||||
NM_001146262.2 | Intron | NP_001139734.1 | ||||
NM_001146264.2 | Intron | NP_001139736.1 | ||||
NM_001256006.1 | Intron | NP_001242935.1 | ||||
NM_153262.3 | Intron | NP_694994.2 | ||||
XM_006711262.3 | Intron | XP_006711325.1 | ||||
XM_017000931.1 | Intron | XP_016856420.1 | ||||
XM_017000932.1 | Intron | XP_016856421.1 | ||||
XM_017000933.1 | Intron | XP_016856422.1 | ||||
XM_017000934.1 | Intron | XP_016856423.1 | ||||
XM_017000935.1 | Intron | XP_016856424.1 |