Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGACCTGTGTTTTTCCCCTGGCT[C/G]CTTTGGAAATTATGGGAAAAAATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601276 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF177 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF177 - zinc finger protein 177 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172651.1 | 177 | Intron | NP_001166122.1 | |||
NM_003451.2 | 177 | UTR 5 | NP_003442.2 |
ZNF559-ZNF177 - ZNF559-ZNF177 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172650.2 | 177 | Intron | NP_001166121.1 | |||
NM_001202425.1 | 177 | UTR 5 | NP_001189354.1 |