Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTTGCTGGGGACCCTCTGCTCC[C/T]GTGTCTTCATGCACCCTCCTACTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109750 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLVRA PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BLVRA - biliverdin reductase A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000712.3 | Intron | NP_000703.2 | ||||
NM_001253823.1 | Intron | NP_001240752.1 | ||||
XM_011515474.2 | Intron | XP_011513776.1 | ||||
XM_017012520.1 | Intron | XP_016868009.1 |
Set Membership: |
HapMap |