Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTATGAGAGCTTTTGACCCAGT[C/T]CTCTCCTAATAGCAGGTGTGTGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZFP2 - ZFP2 zinc finger protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF454 - zinc finger protein 454 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178089.2 | Intron | NP_001171560.1 | ||||
NM_001178090.2 | Intron | NP_001171561.1 | ||||
NM_001323306.1 | Intron | NP_001310235.1 | ||||
NM_001323307.1 | Intron | NP_001310236.1 | ||||
NM_001323308.1 | Intron | NP_001310237.1 | ||||
NM_001323309.1 | Intron | NP_001310238.1 | ||||
NM_001323310.1 | Intron | NP_001310239.1 | ||||
NM_182594.3 | Intron | NP_872400.2 |
Set Membership: |
HapMap |