Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605299 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCOA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCOA6 - nuclear receptor coactivator 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242539.2 | Intron | NP_001229468.1 | ||||
NM_001318240.1 | Intron | NP_001305169.1 | ||||
NM_014071.4 | Intron | NP_054790.2 | ||||
XM_006723755.3 | Intron | XP_006723818.1 | ||||
XM_011528721.2 | Intron | XP_011527023.1 | ||||
XM_011528726.2 | Intron | XP_011527028.1 | ||||
XM_011528727.2 | Intron | XP_011527029.1 | ||||
XM_017027739.1 | Intron | XP_016883228.1 | ||||
XM_017027740.1 | Intron | XP_016883229.1 | ||||
XM_017027741.1 | Intron | XP_016883230.1 | ||||
XM_017027742.1 | Intron | XP_016883231.1 | ||||
XM_017027743.1 | Intron | XP_016883232.1 | ||||
XM_017027744.1 | Intron | XP_016883233.1 | ||||
XM_017027745.1 | Intron | XP_016883234.1 | ||||
XM_017027746.1 | Intron | XP_016883235.1 | ||||
XM_017027747.1 | Intron | XP_016883236.1 | ||||
XM_017027748.1 | Intron | XP_016883237.1 | ||||
XM_017027749.1 | Intron | XP_016883238.1 | ||||
XM_017027750.1 | Intron | XP_016883239.1 | ||||
XM_017027751.1 | Intron | XP_016883240.1 |