Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTGGGTTGAGAAGGGAGAAATA[C/T]TGGTTCATAGAAGACTTCCTATAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602136 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEX1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PEX1 - peroxisomal biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM48 - RNA binding motif protein 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032120.3 | Intron | NP_115496.2 | ||||
XM_005250636.4 | Intron | XP_005250693.1 | ||||
XM_006716149.3 | Intron | XP_006716212.1 | ||||
XM_006716150.3 | Intron | XP_006716213.1 | ||||
XM_017012709.1 | Intron | XP_016868198.1 |