Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCCCTCCTCTGTGACTTCTGTC[A/G]TACATAGCCTGAGTATGTTGGCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609840 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC24A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC24A4 - solute carrier family 24 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153646.3 | Intron | NP_705932.2 | ||||
NM_153647.3 | Intron | NP_705933.2 | ||||
NM_153648.3 | Intron | NP_705934.1 | ||||
XM_005267342.2 | Intron | XP_005267399.1 | ||||
XM_011536436.2 | Intron | XP_011534738.1 | ||||
XM_011536437.2 | Intron | XP_011534739.1 | ||||
XM_011536438.2 | Intron | XP_011534740.1 | ||||
XM_011536439.2 | Intron | XP_011534741.1 | ||||
XM_011536440.2 | Intron | XP_011534742.1 | ||||
XM_011536441.2 | Intron | XP_011534743.1 | ||||
XM_011536442.2 | Intron | XP_011534744.1 |