Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606376 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CHST10 - carbohydrate sulfotransferase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004854.4 | 2551 | UTR 3 | NP_004845.1 | |||
XM_011512207.1 | 2551 | UTR 3 | XP_011510509.1 | |||
XM_011512208.1 | 2551 | UTR 3 | XP_011510510.1 | |||
XM_011512210.1 | 2551 | UTR 3 | XP_011510512.1 | |||
XM_011512211.1 | 2551 | UTR 3 | XP_011510513.1 | |||
XM_011512212.1 | 2551 | UTR 3 | XP_011510514.1 | |||
XM_017005380.1 | 2551 | UTR 3 | XP_016860869.1 | |||
XM_017005381.1 | 2551 | UTR 3 | XP_016860870.1 | |||
XM_017005382.1 | 2551 | UTR 3 | XP_016860871.1 | |||
XM_017005383.1 | 2551 | UTR 3 | XP_016860872.1 |
Set Membership: |
HapMap DME Validated Inventoried |