Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605325 | ||||||||||||||||||||||||||||||||
Literature Links: |
CYP3A5 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
CYP3A5*3J,g.29782A>G | ||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CYP3A5 - cytochrome P450 family 3 subfamily A member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000777.4 | 1996 | Missense Mutation | ATC,GTC | I,V 456 | NP_000768.1 | |
NM_001190484.2 | 1996 | Intron | NP_001177413.1 | |||
NM_001291829.1 | 1996 | Missense Mutation | ATC,GTC | I,V 343 | NP_001278758.1 | |
NM_001291830.1 | 1996 | Missense Mutation | ATC,GTC | I,V 446 | NP_001278759.1 |
ZSCAN25 - zinc finger and SCAN domain containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145115.2 | 1996 | Intron | NP_660090.2 |
Set Membership: |
DME Validated Inventoried |