Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138380 MIM: 138333 | ||||||||||||||||||||||||||||||||
Literature Links: |
GSTM2 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GSTM2 - glutathione S-transferase mu 2 (muscle) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GSTM4 - glutathione S-transferase mu 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000850.4 | 712 | Missense Mutation | AAA,ACA | K,T 133 | NP_000841.1 | |
NM_147148.2 | 712 | Missense Mutation | AAA,ACA | K,T 133 | NP_671489.1 | |
XM_017001085.1 | 712 | Missense Mutation | AAA,ACA | K,T 133 | XP_016856574.1 | |
XM_017001086.1 | 712 | Missense Mutation | AAA,ACA | K,T 92 | XP_016856575.1 | |
XM_017001087.1 | 712 | Missense Mutation | AAA,ACA | K,T 92 | XP_016856576.1 | |
XM_017001088.1 | 712 | Missense Mutation | AAA,ACA | K,T 29 | XP_016856577.1 |
Set Membership: |
DME Validated Inventoried |