Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612477 MIM: 609380 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQCG PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IQCG - IQ motif containing G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LMLN - leishmanolysin like peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136049.2 | Intron | NP_001129521.2 | ||||
NM_033029.3 | Intron | NP_149018.2 | ||||
XM_011513270.1 | Intron | XP_011511572.1 | ||||
XM_011513271.1 | Intron | XP_011511573.1 | ||||
XM_017007428.1 | Intron | XP_016862917.1 |
Set Membership: |
HapMap |