Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACCCATTTCTACACAGACAAAT[G/T]AAAAGTTCTGAGAAATCAGGCTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603001 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2V2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UBE2V2 - ubiquitin conjugating enzyme E2 V2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003350.2 | Intron | NP_003341.1 | ||||
XM_005251300.2 | Intron | XP_005251357.1 | ||||
XM_011517583.2 | Intron | XP_011515885.1 | ||||
XM_017013808.1 | Intron | XP_016869297.1 |
Set Membership: |
HapMap |