Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCCCACCTTCCGTGCTCACGTC[A/G]ACGATGTGCAGCTTGGGCAGGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616315 MIM: 606533 | ||||||||||||||||||||
Literature Links: |
C9orf142 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C9orf142 - chromosome 9 open reading frame 142 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLIC3 - chloride intracellular channel 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004669.2 | 783 | Silent Mutation | GTC,GTT | V,V 183 | NP_004660.2 | |
XM_017015281.1 | 783 | Silent Mutation | GTC,GTT | V,V 183 | XP_016870770.1 | |
XM_017015282.1 | 783 | Silent Mutation | GTC,GTT | V,V 154 | XP_016870771.1 |
LCNL1 - lipocalin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |