Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTGCGAGGCAACCGCGCCGACA[T/G]CCAGGACGCAGGGACCACAGACACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPAN18 - tetraspanin 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130783.4 | Intron | NP_570139.3 | ||||
XM_005253217.3 | Intron | XP_005253274.1 | ||||
XM_006718372.3 | Intron | XP_006718435.1 | ||||
XM_006718373.3 | Intron | XP_006718436.1 | ||||
XM_011520459.2 | Intron | XP_011518761.1 | ||||
XM_017018530.1 | Intron | XP_016874019.1 | ||||
XM_017018531.1 | Intron | XP_016874020.1 |