Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGGGGAAGGAGGCCCTTCCCTGT[A/G]AACAGATAATTGCTGCTACCTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611692 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB34 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZBTB34 - zinc finger and BTB domain containing 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099270.1 | Intron | NP_001092740.1 | ||||
XM_005251989.4 | Intron | XP_005252046.1 | ||||
XM_011518699.2 | Intron | XP_011517001.1 |
Set Membership: |
HapMap |