Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGCTACCTACTGGTCTGTATAA[A/C]AGAGAACTAGCAAACCTGCAGTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613543 MIM: 610012 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLCO5A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLCO5A1 - solute carrier organic anion transporter family member 5A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146008.1 | 7586 | Intron | NP_001139480.1 | |||
NM_001146009.1 | 7586 | Intron | NP_001139481.1 | |||
NM_030958.2 | 7586 | Intron | NP_112220.2 | |||
XM_005251313.2 | 7586 | UTR 3 | XP_005251370.1 | |||
XM_017013883.1 | 7586 | UTR 3 | XP_016869372.1 | |||
XM_017013884.1 | 7586 | Intron | XP_016869373.1 | |||
XM_017013885.1 | 7586 | UTR 3 | XP_016869374.1 | |||
XM_017013886.1 | 7586 | Intron | XP_016869375.1 |
SULF1 - sulfatase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |