Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTTGGTCAGGGAACTATTTTGA[A/G]GGGGAAAAATCAAGATAAAGGCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300182 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXorf38 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CXorf38 - chromosome X open reading frame 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MED14 - mediator complex subunit 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004229.3 | Intron | NP_004220.2 | ||||
XM_005272701.2 | Intron | XP_005272758.1 | ||||
XM_011544000.2 | Intron | XP_011542302.1 | ||||
XM_017029962.1 | Intron | XP_016885451.1 |
Set Membership: |
HapMap |