Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGTGTGATTCTGGGACGACAGC[A/G]TTTTGGGGAGCTACAGGCTTGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
62 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616765 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130417 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100130417 - uncharacterized LOC100130417 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107985728 - collagen alpha-1(III) chain-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC284600 - uncharacterized LOC284600 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAMD11 - sterile alpha motif domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152486.2 | Intron | NP_689699.2 |
Set Membership: |
HapMap |