Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGTTGTTCTTTGGGAAAGCTTA[A/G]GTGAGTTGATTGTTTATTAAGCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608726 | ||||||||||||||||||||
Literature Links: |
PHACTR4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHACTR4 - phosphatase and actin regulator 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048183.1 | Intron | NP_001041648.1 | ||||
NM_023923.3 | Intron | NP_076412.3 | ||||
XM_005245970.2 | Intron | XP_005246027.1 | ||||
XM_017002157.1 | Intron | XP_016857646.1 | ||||
XM_017002158.1 | Intron | XP_016857647.1 | ||||
XM_017002159.1 | Intron | XP_016857648.1 | ||||
XM_017002160.1 | Intron | XP_016857649.1 | ||||
XM_017002161.1 | Intron | XP_016857650.1 | ||||
XM_017002162.1 | Intron | XP_016857651.1 | ||||
XM_017002163.1 | Intron | XP_016857652.1 | ||||
XM_017002164.1 | Intron | XP_016857653.1 | ||||
XM_017002165.1 | Intron | XP_016857654.1 | ||||
XM_017002166.1 | Intron | XP_016857655.1 | ||||
XM_017002167.1 | Intron | XP_016857656.1 | ||||
XM_017002168.1 | Intron | XP_016857657.1 | ||||
XM_017002169.1 | Intron | XP_016857658.1 | ||||
XM_017002170.1 | Intron | XP_016857659.1 | ||||
XM_017002171.1 | Intron | XP_016857660.1 | ||||
XM_017002172.1 | Intron | XP_016857661.1 | ||||
XM_017002173.1 | Intron | XP_016857662.1 |