Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCCACTATGGCAACACTAATTT[A/C]TTGGTAAATGCAAACCACTATAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608726 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHACTR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PHACTR4 - phosphatase and actin regulator 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048183.1 | Intron | NP_001041648.1 | ||||
NM_023923.3 | Intron | NP_076412.3 | ||||
XM_005245970.2 | Intron | XP_005246027.1 | ||||
XM_017002157.1 | Intron | XP_016857646.1 | ||||
XM_017002158.1 | Intron | XP_016857647.1 | ||||
XM_017002159.1 | Intron | XP_016857648.1 | ||||
XM_017002160.1 | Intron | XP_016857649.1 | ||||
XM_017002161.1 | Intron | XP_016857650.1 | ||||
XM_017002162.1 | Intron | XP_016857651.1 | ||||
XM_017002163.1 | Intron | XP_016857652.1 | ||||
XM_017002164.1 | Intron | XP_016857653.1 | ||||
XM_017002165.1 | Intron | XP_016857654.1 | ||||
XM_017002166.1 | Intron | XP_016857655.1 | ||||
XM_017002167.1 | Intron | XP_016857656.1 | ||||
XM_017002168.1 | Intron | XP_016857657.1 | ||||
XM_017002169.1 | Intron | XP_016857658.1 | ||||
XM_017002170.1 | Intron | XP_016857659.1 | ||||
XM_017002171.1 | Intron | XP_016857660.1 | ||||
XM_017002172.1 | Intron | XP_016857661.1 | ||||
XM_017002173.1 | Intron | XP_016857662.1 |
Set Membership: |
HapMap |