Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGAAGAGATGGCTCTCTGGCCA[C/T]GTCCTAACACCAGACTGTCCTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611223 MIM: 613524 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKT3 - AKT serine/threonine kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206729.1 | Intron | NP_001193658.1 | ||||
NM_005465.4 | Intron | NP_005456.1 | ||||
NM_181690.2 | Intron | NP_859029.1 | ||||
XM_005272994.4 | Intron | XP_005273051.1 | ||||
XM_005272995.2 | Intron | XP_005273052.1 | ||||
XM_006711726.3 | Intron | XP_006711789.1 | ||||
XM_011544012.2 | Intron | XP_011542314.1 | ||||
XM_011544013.2 | Intron | XP_011542315.1 | ||||
XM_011544014.2 | Intron | XP_011542316.1 | ||||
XM_016999985.1 | Intron | XP_016855474.1 |
SDCCAG8 - serologically defined colon cancer antigen 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006642.3 | Intron | NP_006633.1 | ||||
XM_005273013.4 | Intron | XP_005273070.1 | ||||
XM_005273018.1 | Intron | XP_005273075.1 | ||||
XM_005273021.4 | Intron | XP_005273078.1 | ||||
XM_005273022.3 | Intron | XP_005273079.1 | ||||
XM_005273023.4 | Intron | XP_005273080.1 | ||||
XM_006711727.3 | Intron | XP_006711790.1 | ||||
XM_006711728.3 | Intron | XP_006711791.1 | ||||
XM_011544021.2 | Intron | XP_011542323.1 | ||||
XM_011544022.2 | Intron | XP_011542324.1 | ||||
XM_011544023.2 | Intron | XP_011542325.1 | ||||
XM_011544024.2 | Intron | XP_011542326.1 | ||||
XM_011544026.2 | Intron | XP_011542328.1 | ||||
XM_011544028.2 | Intron | XP_011542330.1 | ||||
XM_011544029.2 | Intron | XP_011542331.1 | ||||
XM_011544030.2 | Intron | XP_011542332.1 | ||||
XM_017000103.1 | Intron | XP_016855592.1 | ||||
XM_017000104.1 | Intron | XP_016855593.1 | ||||
XM_017000105.1 | Intron | XP_016855594.1 | ||||
XM_017000106.1 | Intron | XP_016855595.1 | ||||
XM_017000107.1 | Intron | XP_016855596.1 | ||||
XM_017000108.1 | Intron | XP_016855597.1 | ||||
XM_017000109.1 | Intron | XP_016855598.1 |
Set Membership: |
HapMap |