Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608662 | ||||||||||||||||||||
Literature Links: |
ANO5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANO5 - anoctamin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142649.1 | Intron | NP_001136121.1 | ||||
NM_213599.2 | Intron | NP_998764.1 | ||||
XM_005252820.3 | Intron | XP_005252877.2 | ||||
XM_005252821.3 | Intron | XP_005252878.2 | ||||
XM_005252822.4 | Intron | XP_005252879.1 | ||||
XM_011519949.2 | Intron | XP_011518251.1 |