Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATATGGTCTCTACGTGTTGGTG[C/T]TGCTATGGGAGAGAGAACCAGGGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605906 MIM: 606665 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDB3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
OPN4 - opsin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030015.2 | 1698 | Intron | NP_001025186.1 | |||
NM_033282.3 | 1698 | Intron | NP_150598.1 | |||
XM_017016955.1 | 1698 | Intron | XP_016872444.1 | |||
XM_017016956.1 | 1698 | Intron | XP_016872445.1 | |||
XM_017016957.1 | 1698 | Intron | XP_016872446.1 |
Set Membership: |
HapMap |