Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCACAGTGATACTTCATTCCAC[G/T]GAGAACCATCCATGGGTGCTGAAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf74 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C8orf74 - chromosome 8 open reading frame 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040032.1 | Intron | NP_001035121.1 | ||||
XM_011543816.2 | Intron | XP_011542118.1 |
MIR4286 - microRNA 4286 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |