Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCAAGATTGGGCAGGAAGGATTG[G/T]GTGGTGGTGAAGAGCGCAGGATATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612974 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEPTOR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DEPTOR - DEP domain containing MTOR-interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283012.1 | Intron | NP_001269941.1 | ||||
NM_022783.3 | Intron | NP_073620.2 | ||||
XM_017013735.1 | Intron | XP_016869224.1 |
Set Membership: |
HapMap |