Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTTCCCTGAGCATTCCCACCTCA[A/G]GTCGTGGCTCTCTGCGGCCCCCCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606594 | ||||||||||||||||||||
Literature Links: |
SETD7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SETD7 - SET domain containing lysine methyltransferase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306199.1 | Intron | NP_001293128.1 | ||||
NM_001306200.1 | Intron | NP_001293129.1 | ||||
NM_030648.3 | Intron | NP_085151.1 | ||||
XM_017008661.1 | Intron | XP_016864150.1 |