Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGATTATTTTAACACTAGAGAG[A/C]TAAATGGCATGATTGTATTGCTACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114815 | ||||||||||||||||||||
Literature Links: |
CA8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CA8 - carbonic anhydrase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321837.1 | Intron | NP_001308766.1 | ||||
NM_001321838.1 | Intron | NP_001308767.1 | ||||
NM_001321839.1 | Intron | NP_001308768.1 | ||||
NM_004056.5 | Intron | NP_004047.3 | ||||
XM_011517587.2 | Intron | XP_011515889.1 | ||||
XM_011517588.2 | Intron | XP_011515890.1 | ||||
XM_017013818.1 | Intron | XP_016869307.1 |