Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606883 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRAK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IRAK4 - interleukin 1 receptor associated kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114182.2 | Intron | NP_001107654.1 | ||||
NM_001145256.1 | Intron | NP_001138728.1 | ||||
NM_001145257.1 | Intron | NP_001138729.1 | ||||
NM_001145258.1 | Intron | NP_001138730.1 | ||||
NM_016123.3 | Intron | NP_057207.2 | ||||
XM_005268943.3 | Intron | XP_005269000.1 | ||||
XM_005268944.4 | Intron | XP_005269001.1 | ||||
XM_005268945.4 | Intron | XP_005269002.1 | ||||
XM_005268947.4 | Intron | XP_005269004.1 | ||||
XM_005268948.2 | Intron | XP_005269005.1 | ||||
XM_005268949.2 | Intron | XP_005269006.1 | ||||
XM_006719438.3 | Intron | XP_006719501.1 | ||||
XM_006719439.2 | Intron | XP_006719502.1 | ||||
XM_006719440.2 | Intron | XP_006719503.1 | ||||
XM_006719441.2 | Intron | XP_006719504.1 | ||||
XM_006719442.2 | Intron | XP_006719505.1 | ||||
XM_006719443.3 | Intron | XP_006719506.1 | ||||
XM_011538431.2 | Intron | XP_011536733.1 | ||||
XM_011538432.1 | Intron | XP_011536734.1 | ||||
XM_011538433.2 | Intron | XP_011536735.1 | ||||
XM_017019390.1 | Intron | XP_016874879.1 | ||||
XM_017019391.1 | Intron | XP_016874880.1 |
PUS7L - pseudouridylate synthase 7 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |