Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTCCATTTGTGGAGCATTTTAT[A/G]GTTTAGAAAGTGCTTTGACATAGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R21 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPP1R21 - protein phosphatase 1 regulatory subunit 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135629.2 | Intron | NP_001129101.1 | ||||
NM_001193475.1 | Intron | NP_001180404.1 | ||||
NM_152994.4 | Intron | NP_694539.1 | ||||
XM_017003301.1 | Intron | XP_016858790.1 | ||||
XM_017003302.1 | Intron | XP_016858791.1 |