Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTCTCTGCAAGGCTGTGGAGCA[C/T]GGACTGACCATCTCTGGGATTGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179590 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRF - protein tyrosine phosphatase, receptor type F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005271079.3 | Intron | XP_005271136.1 | ||||
XM_005271080.3 | Intron | XP_005271137.1 | ||||
XM_005271081.3 | Intron | XP_005271138.1 | ||||
XM_005271082.3 | Intron | XP_005271139.1 | ||||
XM_006710795.3 | Intron | XP_006710858.1 | ||||
XM_006710796.3 | Intron | XP_006710859.1 | ||||
XM_006710797.3 | Intron | XP_006710860.1 | ||||
XM_006710798.3 | Intron | XP_006710861.1 | ||||
XM_006710799.3 | Intron | XP_006710862.1 | ||||
XM_006710800.3 | Intron | XP_006710863.1 | ||||
XM_006710801.3 | Intron | XP_006710864.1 | ||||
XM_011541871.2 | Intron | XP_011540173.1 | ||||
XM_011541872.2 | Intron | XP_011540174.1 | ||||
XM_011541873.2 | Intron | XP_011540175.1 | ||||
XM_017001942.1 | Intron | XP_016857431.1 | ||||
XM_017001943.1 | Intron | XP_016857432.1 | ||||
XM_017001944.1 | Intron | XP_016857433.1 | ||||
XM_017001945.1 | Intron | XP_016857434.1 | ||||
XM_017001946.1 | Intron | XP_016857435.1 | ||||
XM_017001947.1 | Intron | XP_016857436.1 | ||||
XM_017001948.1 | Intron | XP_016857437.1 | ||||
XM_017001949.1 | Intron | XP_016857438.1 | ||||
XM_017001950.1 | Intron | XP_016857439.1 |