Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGGAGGTGGCTTGGCTTGTCTA[C/T]GTGCTGGGCCAACAGCCACTGGCGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALPK3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALPK3 - alpha kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020778.4 | 191 | Silent Mutation | TAC,TAT | Y,Y 8 | NP_065829.3 | |
XM_017022444.1 | 191 | Silent Mutation | TAC,TAT | Y,Y 8 | XP_016877933.1 | |
XM_017022445.1 | 191 | Silent Mutation | TAC,TAT | Y,Y 8 | XP_016877934.1 |